Learn More
ATCC Human rhinovirus 1B; Strain: B632
Supplier:  ATCC VR1645
Encompass
Human rhinovirus 1B strain B632 is propagated in H1HeLa cells (ATCC CRL-1958). This strain was isolated from human throat washings and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in respiratory disease research and infectious disease research.
Catalog No. 50-238-3370