Learn More
Abcam Human RIP3 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Supplier:  Abcam AB28980310X96TESTS
Encompass_Preferred
Human RIP3 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human RIP3.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2311896