Learn More
ACROBiosystems Human ROR1 25tests
Supplier:  ACROBiosystems RO1HA2H425TESTS
Encompass_Preferred
APC-Labeled Human ROR1 Protein His Tag (RO1-HA2H4) is produced via conjugation of APC to Human ROR1 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human ROR1 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).
Catalog No. 50-253-4237