Learn More
Sino Biological Human respiratory syncytial virus (RSV) Glycoprotein G / RSV-G Antibody, Mouse MAb
Supplier:  Sino Biological 11070MM15100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RSV Glycoprotein G/RSV-G (HRSV-G; Catalog#11070-V08H2; Asn66-Arg297; P27022-1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. NC1847841