Learn More
Raybiotech Inc Human SERPINA4 ELISA Kit
Supplier:  Raybiotech Inc ELHSERPINA41
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.12 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Serpin A4. Other species not determined.
Catalog No. 50-194-9713