Learn More
ACROBiosystems ACROBiosystems Human Serpin B9 / SERPINB9 Protein, His Tag
Supplier:  ACROBiosystems SE9H55H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Serpin B9, His Tag (SE9-H55H3) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Pro 376 (Accession # P50453-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7082