Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Human serum albumin antibody (YA985) | 10 UL
SDP

Supplier:  Medchemexpress LLC HYP81281A10UL

Encompass_Preferred

This Human Serum Albumin Antibody (YA985) is a mouse-derived IgG1 monoclonal antibody designed to target human serum albumin. Human serum albumin is the most abundant protein in human blood, crucial for regulating blood plasma colloid osmotic pressure and acting as a carrier for various endogenous molecules and exogenous drugs.

  • Mouse-derived IgG1 monoclonal antibody
  • Non-conjugated
  • Targets human serum albumin
  • Host: Mouse
  • Reactivity: Human
  • Affinity purified
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, pH 7.3
  • Predicted and observed molecular weight: 69 kDa
  • Applications include WB (1:500-1:1000) and ELISA (1:10000)

Catalog No. 50-004-43050


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.