Learn More
Abcam Human Albumin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1798871X96TESTS
Encompass_Preferred
Human Albumin ELISA kit is a single-wash, 90-minute SimpleStep ELISA used to quantify human and cynomolgus monkey albumin (ALB) with a sensitivity of 0.1 ng/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout, compatible with any standard plate reader - Different formats for different needs: -384-well format available for high-throughput applications - Broad sample compatibility – works with serum, plasma (heparin, citrate, EDTA), and cell culture supernatants - Cited in over 15 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1162911