Learn More
Rockland Immunochemicals HUMAN SERUM (Lyophilized) - D314-05
SDP
Supplier:  Rockland Immunochemicals D31405
Encompass
Human Serum is used as a component of bioassays, immunoassays or enzyme assays. Human Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Human serum is also available as a sterile preparation for use as a cell culture supplement.
Catalog No. 50-105-8917