Learn More
MOLECULAR DEPOT LLC Human Serum Proteins - 1 g
Supplier:  MOLECULAR DEPOT LLC K2010001EB1.0
Encompass_Preferred
Human Serum Proteins Powder (1 g) is highly purified human serum proteins provided in powder form This product useful for testing the interference of total serum proteins For in vitro research use only
Catalog No. 50-000-13576