Learn More
Sino Biological Recombinant Human SIGIRR Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 12165H08HB100
Encompass_Preferred
A DNA sequence encoding the Human SIGIRR (Q6IA17-1) (Met1-His118) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-241-2428