Learn More
ACROBiosystems Human Siglec-2 2.5mg
Supplier:  ACROBiosystems MBSC0072.5MG
Encompass_Preferred
ActiveMax Human CD22 uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human CD22 Protein expressed from human 293 cells (HEK293) and contains AA Asp 20 - Arg 687 (Accession # P20273-1).
Catalog No. 50-253-4464