Learn More
ACROBiosystems Alexa Fluor 488-Labeled Human Siglec-2 / CD22 Protein, His TagStar Staining
Supplier: ACROBiosystems SI2HA2H725UG
Alexa Fluor 488-Labeled Human Siglec-2, His Tag (SI2-HA2H7) is produced via conjugation of AF488 to Human Siglec-2, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-2, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1).