Learn More
Bon Opus Biosciences Recombinant Human SIGLEC-8 (C-mFc) 50ug
Supplier: Bon Opus Biosciences BP105450UG
Recombinant Human SIGLEC-8 (C-mFc) is produced by Human Cells. The target gene encoding Met17-Ala363 is expressed with a C-mFc tag. Accession# Q9NYZ4-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1054