Learn More
ACROBiosystems ACROBiosystems Human SIRP beta / CD172b Protein, Fc Tag
Supplier:  ACROBiosystems SIAH5257100UG
Encompass_Preferred
Recombinant Protein;100UG;Human SIRP beta, Fc Tag (SIA-H5257) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Leu 371 (Accession # O00241-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.7 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6706