Learn More
Sino Biological Anti-SLAMF6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11945RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Ly108 / SLAMF6 (rh Ly108 / SLAMF6; Catalog#11945-H08H; Q96DU3-1; Met 1-Met 226). Ly108 / SLAMF6 specific IgG was purified by Human Ly108 / SLAMF6 affinity chromatography.
Catalog No. 50-241-0356