Learn More
Sino Biological Recombinant Human SLC39A6/LIV-1 Protein (ECD, His Tag)
Supplier:  Sino Biological 19372H08H100
Encompass_Preferred
A DNA sequence encoding the Human SLC39A6 (Q13433-1) (Phe29-Trp325) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3280