Learn More
BIOHIPPO Human SNCb Microsample
SDP
Supplier:  BIOHIPPO BHE15210603
Encompass
Human SNCb Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SNCb in Serum plasma cerebrospinal fluid and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SNCb Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SNCb Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SNCb biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15743