Learn More
American Research Products Inc Human soluble fibrin monomer c
Supplier:  American Research Products Inc CSBE09097H
Encompass
Human soluble fibrin monomer compleX SFMC ELISA Kit from CUSABIO Detection Range 6 25 g/mL-400 g/mL Sensitivity 1 56 g/mL Sample serum plasma urine Method Sandwich
Catalog No. 50-003-29556