Learn More
Raybiotech Inc Human ST3GAL1 ELISA Kit
Supplier:  Raybiotech Inc ELHST3GAL11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.8 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human ST3GAL1.
Catalog No. 50-194-9750