Learn More
Sino Biological Human ST6GAL1 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1159015
Encompass_Preferred
The Human ST6GAL1/ST6GAL-I ELISA Pair Set is for the quantitative determination of Human ST6GAL1/ST6GAL-I.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human ST6GAL1/ST6GAL-I coated on a 96-well plate. Standards and samples are added to the wells, and any Human ST6GAL1/ST6GAL-I present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human ST6GAL1/ST6GAL-I polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human ST6GAL1/ST6GAL-I present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86545