Learn More
Sino Biological Anti-PC4/SUB1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14326T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SUB1 / PC4 (rh SUB1 / PC4; Catalog#14326-H07E; P53999; Met1-Leu127). SUB1 / PC4 specific IgG was purified by Human SUB1 / PC4 affinity chromatography.
Catalog No. 50-240-9420