Learn More
ACROBiosystems Human Tau-441 / 2N4R Pre-formed Fibrils Protein, Tag Free
Supplier:  ACROBiosystems TAUH5115100UG
Encompass_Preferred
Human Tau-441 Pre-formed Fibrils, Tag Free (TAU-H5115) is expressed from E.coli cells. It contains AA Met 1 - Leu 441 (Accession # P10636-8).
Catalog No. 50-233-1516