Learn More
American Research Products Inc Human Tetranectin CLEC3B ELI
Supplier:  American Research Products Inc CSBEL005531HU
Encompass
Human Tetranectin (CLEC3B) ELISA kit from CUSABIO Detection Range 1 56 ng/ml- 100 ng/ml Sensitivity 0 39 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27306