Learn More
Raybiotech Inc HUMAN TGF-BETA 1 ELISA
Supplier:  Raybiotech Inc ELHTGFB11
Encompass_Preferred
RayBio Human TGF beta 1 ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 80 pg/ml - 60 ng/ml
Catalog No. 50-104-4372