Learn More
Raybiotech Inc C-Series Human TGF beta Array C2 Kit.
Supplier:  Raybiotech Inc AAHTGFB24
Encompass_Preferred
Detects 25 proteins in human samples. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Activin A, Activin R1A (ALK-2), BAMBI (NMA), beta IG-H3, BMP-2, BMP-4, BMP-6, BMP-7, BMP-8, BMP-9, BMP-15 (GDF-9B), BMPR-IA (ALK-3), BMPR-II, Follistatin, GDF-11 (BMP-11), GDF-15, Lefty-A, Noggin, SMAD3, Smad4, TGF alpha, TGF beta 1, TGF beta 2, TGF beta R2, TGF beta R3
Catalog No. 50-194-8664