Learn More
ACROBiosystems Human TGF-beta RII / TGFBR2 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems TG2H52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;Human TGF-beta RII, His Tag (TG2-H52H5) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # P37173-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.4 kDa. The protein migrates as 25-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Serial dilutions of Human TGF-beta RII, His Tag (Cat. No. TG2-H52H5) were added into Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) : Biotinylated Human TGF-beta RII, His,Avitag (Cat. No. TG2-H82E4) binding reactions. The half maximal inhibitory concentration (IC50) is 0.39287 μg/mL (QC tested).
Catalog No. 50-205-5077