Learn More
Sino Biological Human TIM-1/HAVCR1 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1105115
Encompass_Preferred
The Human TIM-1/KIM-1/HACVR ELISA Pair Set is for the quantitative determination of Human TIM-1/KIM-1/HACVR.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human TIM-1/KIM-1/HACVR coated on a 96-well plate. Standards and samples are added to the wells, and any Human TIM-1/KIM-1/HACVR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human TIM-1/KIM-1/HACVR monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human TIM-1/KIM-1/HACVR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85197