Learn More
ACROBiosystems Human TIM-3 25ug
Supplier:  ACROBiosystems TI3H82F825UG
Encompass_Preferred
Biotinylated Human TIM-3 Protein FcAvitag (TI3-H82F8) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0-1).
Catalog No. 50-253-4057