Learn More
Biomatik Corporation Human Tissue Factor Pathway Inhibitor (TFPI) CLIA Kit
Supplier:  Biomatik Corporation EKU09456
Encompass_Preferred
Human Tissue Factor Pathway Inhibitor (TFPI) CLIA Kit for detecting TFPI in plasma, urine, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.05-40ng/ml. Sensitivity: 0.03ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P10646. Gene ID: 7035. Target Synonyms: TFI; EPI; LACI; TFPI1; Lipoprotein-Associated Coagulation Inhibitor; Extrinsic pathway inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8517