Learn More
Invivogen Human TLR9 Agonist Kit
SDP
Supplier:  Invivogen TLRLKIT9H
Encompass
This kit features the most widely known CpG ODNs from Class A, B or C that function best with human cells.
Catalog No. NC1333950