Learn More
ACROBiosystems Human TNF-alpha Protein, Fc Tag
Supplier:  ACROBiosystems TNAH5255100UG
Encompass_Preferred
Human TNF-alpha Protein, Fc Tag (TNA-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P01375-1).
Catalog No. 50-233-1571