Learn More
Sino Biological Human TNFR1/CD120a Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK108725
Encompass_Preferred
The human TNFRI / CD120a ELISA Pair Set is for the quantitative determination of human TNFRI.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for TNFRI /CD120a coated on a 96-well plate. Standards and samples are added to the wells,and any TNFRI / CD120a present binds to the immobilized antibody. The wells arewashed and a horseradish peroxidase conjugated mouse anti-TNFRI / CD120amonoclonal antibody is then added, producing an antibody-antigen-antibody"sandwich". The wells are again washed and TMB substrate solution is loaded,which produces color in proportion to the amount of TNFRI / CD120a present inthe sample. To end the enzyme reaction, the stop solution is added andabsorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85104