Learn More
Sino Biological Recombinant Human HVEM Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10334H49HB20
Encompass_Preferred
A DNA sequence encoding the Human TNFRSF14 (NP_003811.2)(Met1-Val202) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1000