Learn More
Sino Biological Recombinant Human BCMA Protein (Site-Specific APC-Conjugated, ECD, His Tag)
Supplier:  Sino Biological 10620H86HSA25
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ala 54) of human TNFRSF17 (NP_001183.2) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with APC (Excitation Max.= 651 nm, Emission Max.= 662 nm).
Catalog No. 50-299-2900