Learn More
Cusabio Technology LLC HUMAN TNFRSF17 ELISA KIT
Supplier:  Cusabio Technology LLC CSBEL023974HU
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Human Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) in serum plasma and tissue homogenates using a sandwich assay format The kit exhibits a detection range of 7 8 pg/ml to 500 pg/ml with a sensitivity of 1 95 pg/ml ensuring accurate measurement across low and high analyte concentrations The assay requires a sample volume of 50-100 l and typically completes within 1-5 hours depending on protocol optimization Absorbance is measured at 450 nm and the standardized workflow ensures reliable results for research or diagnostic applications This kit detects TNFRSF17 with high specificity making it suitable for studies involving immune regulation oncology or related biomarker discovery
Catalog No. 50-000-51667