Learn More
Abcam Human TNFRSF17 ELISA Kit (BCMA), 1 x 96 Tests
Supplier:  Abcam AB2138401X96TESTS
Encompass_Preferred
Human TNFRSF17 ELISA Kit (BCMA) is a Sandwich (quantitative) ELISA kit for the measurement of Human TNFRSF17 (BCMA) in Human in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum, Tissue Homogenate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1690530