Learn More
BIOHIPPO Human TNFRSF1B Microsample
SDP
Supplier:  BIOHIPPO BHE15212256
Encompass
Human TNFRSF1B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TNFRSF1B in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TNFRSF1B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TNFRSF1B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TNFRSF1B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15935