Learn More
Sino Biological Recombinant Human APRIL/TNFSF13 Protein (hFc Tag), MALS-verified
Supplier:  Sino Biological 10610H01H2100
Encompass_Preferred
A DNA sequence encoding the Human TNFSF13 (O75888-1) (Gln111-Leu250) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-299-2869