Learn More
Sino Biological Recombinant Human TL1A/TNFSF15 Protein (MALS-verified, His Tag), MALS-verified
Supplier:  Sino Biological 17049H07H2100
Encompass_Preferred
A DNA sequence encoding the Human TNFSF15 (NP_005109.2) (Leu72-Leu251) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.
Catalog No. 50-299-3523