Learn More
Sino Biological Recombinant Human TL1A/TNFSF15 Protein (His Tag), Trimer, MALS-verified
Supplier:  Sino Biological 17049H07H4100
Encompass_Preferred
A DNA sequence encoding the Human TNFSF15 (NP_005109.2) (Leu72-Leu251) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-3585