Learn More
ACROBiosystems ACROBiosystems Human TRAIL R1 / DR4 / TNFRSF10A Protein, His Tag
Supplier:  ACROBiosystems TR1H5228100UG
Encompass_Preferred
Recombinant Protein;100UG;Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession # O00220-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.8 kDa. The protein migrates as 20-23 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human TRAIL R1, His Tag (Cat. No. TR1-H5228) at 5 μg/mL (100 μL/well) can bind Human TRAIL, Mouse IgG2a Fc Tag (Cat. No. TRL-H5259) with a linear range of 2-63 ng/mL (QC tested).
Catalog No. 50-210-6757