Learn More
BIOHIPPO Human TSC22 Microsample
SDP
Supplier:  BIOHIPPO BHE15212110
Encompass
Human TSC22 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSC22 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 104 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TSC22 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TSC22 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TSC22 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14205