Learn More
Bioworld HUMAN TUMOR NECROSIS FACTOR
Supplier:  Bioworld 220607381
Encompass_Preferred
HUMAN TUMOR NECROSIS FACTOR RECEPTOR FUSION PROTEIN (RECOMBINANT): Recombinant Human Tumor Necrosis Factor Receptor Fusion Protein (TNFR) Source: Chinese Hamster Ovarian Cells (CHO) Background: TNFR binds specifically to tumor necros...
Catalog No. 50-253-836