Learn More
American Research Products Inc Human Tumor susceptibility gen
Supplier:  American Research Products Inc CSBEL025125HU
Encompass
Human Tumor susceptibility gene 101 protein (TSG101) ELISA kit from CUSABIO Detection Range 23 44 pg/mL-1500 pg/mL Sensitivity 5 86 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27631