Learn More
TRIBIOSCIENCE INC Human UCHL1/PGP9.5 ELISA Kit, 1 x 96-well plate
SDP
Supplier:  TRIBIOSCIENCE INC TBS32107
Encompass_Preferred
The Human UCHL1 ELISA is designed to quantitatively detect human UCHL1 levels in CSF, brain or spinal tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 39 to 2500 pg/mL.
Catalog No. NC2886050