Learn More
Sino Biological Human VCAM1 ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological SEK1011315
Encompass_Preferred
Each vial contains 248 ng of recombinant Human VCAM1. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3000 pg/mL is recommended.
Catalog No. 50-237-3262