Learn More
Sino Biological Recombinant Human Vimentin Protein (His Tag)
Supplier:  Sino Biological 10028H08B1
Encompass_Preferred
A DNA sequence encoding the human VIM (Met1-Glu466) (P08670) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-240-7780