Learn More
American Research Products Inc Human Vitamin B12 VB12 ELISA
Supplier:  American Research Products Inc CSBE07903H
Encompass
Human Vitamin B12 VB12 ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 1 498 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26853