Learn More
Eurofins Discoverx Humanin, HN, 1 mg. (92-1221)
Supplier:  Eurofins Discoverx 921221
Encompass
Humanin is a variable length peptide that has neuroprotective, cytoprotective, and anti-apoptotic effects. Mitochondrial derived forms are 21 amino acids, while cytosolic forms are 24 amino acids. Humanin may be useful in protecting against Alzheimer's disease, Huntington's disease, prion disease, stroke, oxidative stress, atherosclerosis, type I diabetes, and heart attack. Acts as an agonist for FPR2 and FPR3. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9744